Learn More
H-2Dk Antibody (15-5-5S), FITC, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP128194
Catalog No. NBP128194
Only null left
Add to Cart
Description
Description
H-2Dk Monoclonal specifically detects H-2Dk in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
H-2Dk
Monoclonal
0.5mg/mL
Flow Cytometry 1 ug/10^6 cells
Mouse
Protein A or G purified
RUO
An epitope on mouse H-2Dk MHC class I alloantigen
Store at 4C. Do not freeze.
IgG2a κ
Flow Cytometry
15-5-5S
FITC
H2Dk, MHC class I
C3H mouse splenocytes
0.25 mg
Primary
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only