Learn More
H-2K[d] Mouse anti-Mouse, FITC, Clone: SF1-1.1, BD
Mouse Monoclonal Antibody
$132.00 - $410.00
Specifications
Antigen | H-2K[d] |
---|---|
Clone | SF1-1.1 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The SF1-1.1 antibody reacts with the α3 domain of the H-2K[d] MHC class I alloantigen. Reactivity with other haplotypes (e.g, b, j, k, p, q, s, v) has not been observed. It has been reported that plate-bound SF1-1.1 mAb moderately enhances the apoptotic response of thymocytes to plate-bound 145-2C11 mAb (anti-mouse CD3e, Cat. No. 557306/553058).
Specifications
H-2K[d] | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Mouse | |
BALB/c mouse cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
SF1-1.1 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.