Learn More
Invitrogen™ H-2Kb/H-2Db Monoclonal Antibody (5041.16.1), PE

Description
MA5-18000 targets H-2KbDb in FACS applications and shows reactivity with Mouse samples. The MA5-18000 immunogen is mouse H-2KbDb.
Specifications
Specifications
| Antigen | H-2Kb/H-2Db |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 5041.16.1 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 4-5mg/mL BSA and 0.02% sodium azide |
| Gene | H2-K1 |
| Gene Accession No. | P01901, P03991, P04223, P14428 |
| Gene Alias | class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H2DB; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2-K1; H2KB; histocompatibility 2, K1, K region; K-f; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.