Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2Kb Mouse anti-Mouse, FITC, Clone: AF6-88.5, BD

Mouse Monoclonal Antibody

$128.00 - $372.00

Specifications

Antigen H-2Kb
Clone AF6-88.5
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB562002
View Documents Promotion Details
BD Biosciences
562002
50 μg
Each for $128.00
Only null left
Add to Cart
 
BDB553569
View Documents Promotion Details
BD Biosciences
553569
0.5 mg
Each for $372.00
Only null left
Add to Cart
 
Description

Description

The AF6-88.5 antibody reacts with the H-2Kb MHC class I alloantigen. Reactivity with other haplotypes (e.g., d, f, j, k, p, q, r, s, u, v) has not been observed.

Specifications

Specifications

H-2Kb
0.5mg/mL
Monoclonal
Mouse
Mouse
Mouse C57BL splenocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
AF6-88.5
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.