Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-2Kb/SIINFEKL Mouse anti-Mouse, PE, Clone: 25-D1.16.rMAb (also known as 25-D1.16), BD Phrmingen™
Mouse Monoclonal Antibody
$110.50 - $333.50
Specifications
Antigen | H-2Kb/SIINFEKL |
---|---|
Clone | 25-D1.16.rMAb (also known as 25-D1.16) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
H-2Kb/SIINFEKL Monoclonal antibody specifically detects H-2Kb/SIINFEKL in Mouse samples. It is validated for Flow cytometrySpecifications
H-2Kb/SIINFEKL | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
14972 | |
IgG1 κ | |
Affinity Chromatography |
25-D1.16.rMAb (also known as 25-D1.16) | |
Flow Cytometry | |
PE | |
Mouse | |
Mouse | |
H-2Kb/SIINFEKL; H-2Kb:SIINFEKL; SIINFEKL-bound H-2Kb | |
SIINFEKL-pulsed RMA-S cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title