Learn More
Invitrogen™ H-2Kd Monoclonal Antibody (31-3-4S)

Description
MA1-70107 detects a private determinant (H-2.31) of the H-2Kd antigen in mouse samples. MA1-70107 has been successfully used in flow cytometry procedures. The MA1-70107 immunogen is H-2Kd from BALB/c.
Specifications
Specifications
| Antigen | H-2Kd |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 31-3-4S |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | H2-K1 |
| Gene Accession No. | P01901, P03991, P04223, P14428 |
| Gene Alias | class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2-K1; histocompatibility 2, K1, K region; K-f; MGC184092; MGC7052; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.