Learn More
H-2Kd Mouse anti-Mouse, Brilliant Violet 421, Clone: SF1-1.1, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 742431
Description
The SF1-1.1 antibody reacts with the 3 domain of the H-2K[d] MHC class I alloantigen. Reactivity with other haplotypes (e.g, b, j, k, p, q, s, v ) has not been observed. It has been reported that plate-bound SF1-1.1 mAb moderately enhances the apoptotic response of thymocytes to plate-bound 145-2C11 mAb (anti-mouse CD3e).
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
H-2Kd | |
SF1-1.1 | |
Brilliant Violet 421 | |
H-2kd, H2kd, MHC class I H-2Kd | |
BALB/c Mouse Cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.