Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-2Kq Mouse anti-Mouse, Brilliant Violet 510, Clone: KH114, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 742297
Description
The KH114 antibody specifically recognizes the H-2K[q] MHC class I alloantigen. Reactivity with other haplotypes (e.g., a, b, d, f, k, n, p, r, s, u, v /) has not been observed.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
H-2Kq | |
KH114 | |
Brilliant Violet 510 | |
H-2Kq, H2-Kq, Histocompatibility-2Kq | |
B10.Q mouse skin graft and splenocytes | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction