Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H-2Ld Monoclonal Antibody (30-5-7S), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518006
Description
MA5-18006 targets H-2Ld in CT and FACS applications and shows reactivity with Mouse samples. The MA5-18006 immunogen is mouse H-2Ld (public determinant H-2.65).
MHC Class I (H-2Ld) is a protein complex located on the surface of cells in vertebrates. Its main function is to present peptides derived from intracellular proteins to T cells for immune recognition. The structure of MHC Class I (H-2Ld) consists of a heavy chain, beta-2 microglobulin, and a peptide-binding groove that interacts with antigenic peptides. The location of MHC Class I (H-2Ld) varies depending on the cell type, but it is typically found on the surface of nucleated cells. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
H-2Ld | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P01897 | |
H2-L | |
mouse H-2Ld (public determinant H-2.65). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
30-5-7S | |
FITC | |
H2-L | |
H2L; H-2L; H2-L; H2LD; histocompatibility 2, D region locus L; histocompatibility 2, L region | |
Mouse | |
Protein G | |
RUO | |
14980 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction