Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H-2Ld Monoclonal Antibody (30-5-7S)
GREENER_CHOICE

Catalog No. PIMA170109
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA170109 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA170109 Supplier Invitrogen™ Supplier No. MA170109
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-70109 detects H-2Ld in mouse samples. MA1-70109 has been successfully used in flow cytometry procedures. The MA1-70109 immunogen is H-2Ld. Stable at 4°C. For long term storage, aliquot and store at -20°C in volumes appropriate for single usage. Avoid freeze/thaw cycles.

MHC Class I (H-2Ld) is a protein complex located on the surface of cells in vertebrates. Its main function is to present peptides derived from intracellular proteins to T cells for immune recognition. The structure of MHC Class I (H-2Ld) consists of a heavy chain, beta-2 microglobulin, and a peptide-binding groove that interacts with antigenic peptides. The location of MHC Class I (H-2Ld) varies depending on the cell type, but it is typically found on the surface of nucleated cells. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H-2Ld
Applications Cytotoxicity Assay, Flow Cytometry
Classification Monoclonal
Clone 30-5-7S
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene H2-L
Gene Accession No. P01897
Gene Alias H2L; H-2L; H2-L; H2LD; histocompatibility 2, D region locus L; histocompatibility 2, L region
Gene Symbols H2-L
Host Species Mouse
Immunogen H-2Ld.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14980
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.