Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ H 89 dihydrochloride
GREENER_CHOICESDP

Catalog No. 291050 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-105-0 50 mg
29-101 1 mg
29-101-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 29-105-0 Supplier R&D Systems™ Supplier No. 2910/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Protein kinase A inhibitor

H 89 dihydrochloride is a protein kinase A inhibitor that also inhibits several other kinases (IC50 values are 80, 120, 135, 270, 2600 and 2800 nM for S6K1, MSK1, PKA, ROCKII, PKBα and MAPKAP-K1b). Exhibits antinociceptive activity. Enhances survival and clonogenicity of dissociated human ESCs through ROCK inhibition.

Chemical Identifiers

CAS 130964-39-5
Molecular Formula C20H24BrCl2N3O3S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 519.28
Purity >99%
Molecular Weight (g/mol) 537.294
Chemical Name or Material H 89 dihydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.