Learn More
H-B Instrument™ Durac™ Thermometer-Hygrometer-Barometers
Recalibrateable instrument
Specifications
Material Stainless steel and brass
View More Specs
Products 2
Catalog Number Mfr. No. Temperature Range (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Temperature Range (Metric) Price Quantity & Availability  
13-201-842
View Documents
H-B Instrument™
B615020100
-20°F to +120°F N/A N/A N/A
13-201-841
View Documents
H-B Instrument™
B615020200
-30°C to +50°C N/A N/A N/A
Description
Description
  • Triple scale barometer feature readings in cmHg, in.Hg and mbar: 940/1058 (mbar), 27.75/31.25 (in.Hg), 70.5 to 79.4 (cmHg)
  • Single scale thermometer and humidity readings with ranges 0 to 100%
  • Stainless-steel and brass construction
  • Hole for wall mounting present
  • Adjust automatically once set
  • Designed to work in altitudes below 6561 ft. (2000m)
Specifications
Specifications
Stainless steel and brass
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.