Learn More
Invitrogen™ H-cadherin Polyclonal Antibody

Description
A suggested positive control is NIH-3T3 cell lysate. PA5-20158 can be used with blocking peptide PEP-0277.
Specifications
Specifications
| Antigen | H-cadherin |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CDH13 |
| Gene Accession No. | P55290, Q9WTR5 |
| Gene Alias | 4932416G01Rik; CAD13; cadherin 13; cadherin 13, H-cadherin (heart); cadherin-13; CD39 antigen-like 3; CD39L3; CD39-like 3; CDH13; CDHH; Cdht; ecto-apyrase 3; ecto-ATPase 3; ecto-ATPDase 3; HB6; H-cadherin; H-cadherin (heart); heart cadherin; heart-cadherin; NTPDase 3; NTPDase-3; P105; T cad; T Cadherin; Tcad; T-cad; T-cadherin; Truncated cadherin; truncated-cadherin |
| Gene Symbols | CDH13 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.