Learn More
R&D Systems™ H/M Pluripotent Stem Cell Multi-Color Flow Cytometry Kit
Click to view available options
Quantity:
25 Tests
Description
Identifying the sources of experimental variability is an important consideration in stem cell research where experiments are costly and time-consuming. A potential source of significant variability arises from the starting population of stem cells which can undergo phenotypic changes in culture.
Defines starting population to reduce variability
Simultaneously verifies stem cell pluripotency using four markers of pluripotency
Does not require the use of secondary antibodies
Specifications
Specifications
Quantity | 25 Tests |
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |