Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H-Ras Recombinant Rabbit Monoclonal Antibody (4Q7M5)
GREENER_CHOICE

Catalog No. PIMA535323
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535323 Supplier Invitrogen™ Supplier No. MA535323
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

H-Ras Recombinant Monoclonal Antibody for ELISA, Western Blot

Monomeric G protein Ras functions as a molecular switch linking receptor and nonreceptor tyrosine kinase activation to a number of downstream signaling pathways leading to distinct cytoplasmic or nuclear events. Each mammalian cell contains three Ras proto-oncogene coding for closely related Ras proteins: H-, K-, N-Ras. Oncogenic mutations in Ras genes are present in ~30% of all human cancers. Constitutive activation of Ras due to mutations or overexpression stimulates proliferation and inhibition of apoptosis. K-Ras mutations are common in pancreatic, colorectal and nonsmall-cell lung carcinomas; H-Ras mutations are common in bladder, kidney and thyroid carcinomas; N-Ras mutations are found in melanoma, hematological malignancies and hepatocellular carcinaoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H-Ras
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4Q7M5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene HRAS
Gene Accession No. P01112, P20171, Q61411
Gene Alias C-BAS/HAS; c-Ha-ras; c-Ha-ras p21 protein; c-Ha-ras transgene; C-HA-RAS1; c-has/bas p21 protein; c-H-ras; c-rasHa; c-ras-Ki-2 activated oncogene; CTLO; GTP- and GDP-binding peptide B; GTPase HRas; GTPase HRas, N-terminally processed; GTPase Hras1; HAMSV; Ha-ras; Ha-Ras1 proto-oncoprotein; Harvey ras1 protein; Harvey rat sarcoma viral (v-Ha-ras) oncogene homolog; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene; Harvey rat sarcoma virus oncogene 1; Harvey-ras; Hras; H-ras; H-ras 1 protein; HRas proto-oncogene, GTPase; HRAS1; H-Ras-1; Hras-1; H-RASIDX; Kras2; p19 H-RasIDX protein; p21ras; ras; Ras family small GTP binding protein H-Ras; ras p21; RASH1; transformation gene: oncogene HAMSV; transforming protein P21; v-Ha-ras Harvey rat sarcoma viral oncogene homolog
Gene Symbols HRAS
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15461, 293621, 3265
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.