Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H1F0 Antibody (1C7), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | H1F0 |
---|---|
Clone | 1C7 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
H1F0 Monoclonal antibody specifically detects H1F0 in Human samples. It is validated for ELISASpecifications
H1F0 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
H1 histone family, member 0, H1.0, H1(0), H1-0, H10, H1FVMGC5241, Histone H1', Histone H1(0), histone H1.0 | |
Synthesized peptide [UniProt P07305] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
1C7 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cellular Markers, Chromatin Research, Epigenetics, Nuclear Envelope Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3005 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title