Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H1F0 Antibody (HH1/1784R), Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50 - $562.50
Specifications
Antigen | H1F0 |
---|---|
Clone | HH1/1784R |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25314420
![]() |
Novus Biologicals
NBP25314420UG |
20 μg |
Each for $304.50
|
|
|||||
NB253144100
![]() |
Novus Biologicals
NBP253144100UG |
100 μg |
Each for $562.50
|
|
|||||
NB253144200
![]() |
Novus Biologicals
NBP253144200UG |
200 μg | N/A | N/A | N/A | ||||
Description
H1F0 Monoclonal specifically detects H1F0 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
H1F0 | |
0.2 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
H1 histone family, member 0, H1.0, H1(0), H1-0, H10, H1FVMGC5241, Histone H1', Histone H1(0), histone H1.0 | |
H1F0 | |
IgG κ | |
Protein A purified | |
30 kDa |
HH1/1784R | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
1mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
3005 | |
Recombinant full-length human Histone H1 protein | |
Primary | |
Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title