Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H1F0 Antibody (r1415-1), Novus Biologicals™

Mouse Monoclonal Antibody
$225.07 - $517.67
Specifications
| Antigen | H1F0 |
|---|---|
| Clone | r1415-1 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25314320
![]() |
Novus Biologicals
NBP25314320UG |
20 μg |
Each for $225.07
|
|
|||||
NB253143100
![]() |
Novus Biologicals
NBP253143100UG |
100 μg |
Each for $517.67
|
|
|||||
NB253143200
![]() |
Novus Biologicals
NBP253143200UG |
200 μg | N/A | N/A | N/A | ||||
Description
H1F0 Monoclonal specifically detects H1F0 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| H1F0 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| H1 histone family, member 0, H1.0, H1(0), H1-0, H10, H1FVMGC5241, Histone H1', Histone H1(0), histone H1.0 | |
| H1F0 | |
| IgG2a κ | |
| Protein A or G purified | |
| 30 kDa |
| r1415-1 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 1mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 3005 | |
| Nuclei of human leukemia biopsy cells | |
| Primary | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title