Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H1F0 Mouse anti-Human, Mouse, Rat, Clone-r1415-1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254303200UG
Description
IHC-P:Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer pH 6.0 for 10-20 min followed by cooling at RT for 20 minutesSpecifications
| H1F0 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| H1 histone family, member 0, H1.0, H1(0), H1-0, H10, H1FVMGC5241, Histone H1', Histone H1(0), histone H1.0 | |
| Mouse | |
| 30 kDa | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| r1415-1 | |
| Unconjugated | |
| PBS with No Preservative | |
| H1F0 | |
| Nuclei of human leukemia biopsy cells | |
| Protein A or G purified | |
| RUO | |
| 3005 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction