Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H1F0 Rabbit anti-Human, Mouse, Rat, Clone: AE-4, Novus Biologicals™
SDP

Catalog No. p-7234231 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122898 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122898 Supplier Novus Biologicals Supplier No. NBP30771220UG
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

H1F0 Monoclonal specifically detects H1F0 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen H1F0
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AE-4
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Formulation 10 mM PBS with 0.05% BSA
Gene Alias H1 histone family, member 0, H1.0, H1(0), H1-0, H10, H1FVMGC5241, Histone H1', Histone H1(0), histone H1.0
Host Species Rabbit
Immunogen Nuclei of human leukemia biopsy cells
Purification Method Protein A purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3005
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.