Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H1FNT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534857 Supplier Invitrogen™ Supplier No. PA534857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Essential for normal spermatogenesis and male fertility. Required for proper cell restructuring and DNA condensation during the elongation phase of spermiogenesis. Involved in the histone-protamine transition of sperm chromatin and the subsequent production of functional sperm. Binds both double-stranded and single-stranded DNA, ATP and protamine-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H1FNT
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene H1FNT
Gene Accession No. Q75WM6
Gene Alias H1 histone family member N, testis specific; H1 histone family, member N, testis-specific; H1.7; H1FNT; H1T2; HANP1; haploid germ cell-specific nuclear protein 1; Histone H1t2; testis secretory sperm-binding protein Li 223n; testis-specific H1 histone
Gene Symbols H1FNT
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Testis-specific H1 histone. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341567
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.