Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H1FOO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558200 Supplier Invitrogen™ Supplier No. PA558200
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YRKTKAESKS SKPTASKVKN GAASPTKKKV VAKAKAPKAG QGPNTKAAAP AKGSGSKVVP AHLSRKTEAP KGPRKAGLPI KASSSKVSSQ Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 36%.

Eukaryotic Histones are basic and water soluble nuclear proteins that form hetero-octameric nucleosome particles by wrapping 146 base pairs of DNA in a left-handed super-helical turn sequentially to form chromosomal fibers. Two molecules of each of the four core Histones (Histone H2A, H2B, H3, and H4) form the octamer, which consists of two H2A-H2B dimers and two H3-H4 dimers that are nearly symmetrical by tertiary structure. Over 80% of nucleosomes contain the linker Histone H1, derived from an intronless gene, that interacts with linker DNA between nucleosomes and mediates compaction into higher order chromatin. H1FOO (H1 histone family, member O, oocyte-specific), also known as OSH1, is a 346 amino acid oocyte-specific Histone that localizes to both the nucleus and the cytoplasm. Expressed as multiple alternatively spliced isoforms, H1FOO is thought to play an important role in gene control during oogenesis and early embryogenesis and is crucial for meiotic maturation of germinal vesicle-stage oocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H1FOO
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene H1FOO
Gene Accession No. Q8IZA3
Gene Alias C86609; H1 histone family member O, oocyte specific; H1 histone family, member O, oocyte-specific; H1.8; H1fo; H1foo; H1OO; Histone H1oo; oocyte-specific histone H1; Oocyte-specific linker histone H1; OSH1; RGD1566236
Gene Symbols H1FOO
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human H1FOO. Recombinant protein control fragment (Product #RP-96499).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132243
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.