Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H1N1 Neuraminidase Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2411100.025MG
Description
H1N1 Neuraminidase Polyclonal specifically detects H1N1 Neuraminidase in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.Specifications
H1N1 Neuraminidase | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Antibody was raised against synthetic peptides from the seasonal influenza (H1N1) Neuraminidase protein and the novel swine influenza Neuraminidase protein. The antibody recognizes both variants. The immunogen is located within the first 50 amino acids of H1N1 Neuraminidase. Amino Acid Squence: SIQLGNQNQIET | |
0.025 mg | |
Primary | |
Influenza virus, Virus | |
IgG |
ELISA | |
1 mg/mL | |
ELISA 1 μg/mL | |
Rabbit | |
Affinity Purified | |
RUO | |
This antibody will recognize the Neuraminidase from the both the seasonal influenza (H1N1) and swine-origin influenza virus (A/California/14/2009 (H1N1)). | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction