Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | H2A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
H2A Polyclonal specifically detects H2A in Human samples. It is validated for Western Blot.Specifications
H2A | |
Polyclonal | |
Purified | |
RUO | |
H2A histone family, member O, H2A.2, H2A/O, H2A/q, H2a-615, H2AFOHistone H2A/o, HIST2H2AAH2A, histone 2, H2aa, histone 2, H2aa3, histone cluster 2, H2aa3, histone H2A type 2-A, Histone H2A.2 | |
HIST2H2AA3 | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Q7L7L0 | |
8337 | |
Synthetic peptides corresponding to HIST2H2AA3(histone cluster 2, H2aa3) The peptide sequence was selected from the middle region of HIST2H2AA3. Peptide sequence PRHLQLAIRNDEELNKLLGKVTIAQGGVLPNIQAVLLPKKTESHHKAKGK. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title