Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2A (H4R3me2s) Mouse anti-Human, Clone: 6G1/1D3, MilliporeSigma™

Catalog No. MABE364MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE364MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE364MI Supplier MilliporeSigma Supplier No. MABE364
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-H2A (H4R3me2s), clone 6G1/1D3 is a highly specific mouse monoclonal antibody that targets H2A/H4R3me2s and has been tested in Dot Blot and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H2A (H4R3me2s)
Applications Dot Blot, Western Blot
Classification Monoclonal
Clone 6G1/1D3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols HIST1H2AG,HIST1H2AG,H2AFP,HIST1H2AI,H2AFC
Host Species Mouse
Immunogen KLH-conjugated linear symmetric di methyl R3 peptide.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003500
Test Specificity Clone 6G1/1D3 specifically detects H2A/H4 arginine 3 symmetric dimethylation (H2A/H4R3me2) in acid extract of HeLa cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.