Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H2A.Zac pan-acetyl (K4,K7,K11,K13) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114690 Supplier Invitrogen™ Supplier No. PA5114690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Acetyl-H2A.Z only when acetylated at Lys4/7/11/13.

H2A.Zac pan-acetyl (K4,K7,K11,K13) is classified under histone H2A which replaces conventional H2A in a subset of nucleosomes. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. May be involved in the formation of constitutive heterochromatin. May be required for chromosome segregation during cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H2A.Zac pan-acetyl (K4,K7,K11,K13)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene H2AFZ
Gene Accession No. P0C0S5
Gene Alias H2A histone family member Z; H2A histone family, member Z; H2A.z; H2a.z-1; H2A/z; H2AFZ; H2az; H2AZ histone; histone H2A.Z
Gene Symbols H2AFZ
Host Species Rabbit
Immunogen A synthesized peptide derived from human H2AFZ(Accession P0C0S5), corresponding to amino acid residues M1-S21.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3015
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.