Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H2AFX, Rabbit anti-Human, FITC, Clone: 1B3, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB18895
Specifications
H2AFX | |
Monoclonal | |
FITC | |
In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
H2A.X/H2A/X/H2AX | |
Rabbit | |
100 Reactions | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
1B3 | |
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user. | |
H2A histone family, member X | |
H2AFX | |
A synthetic phosphopeptide corresponding to residues surrounding S139 of human H2AFX. | |
RUO | |
3014 | |
Store at 4°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction