Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H2AFZ, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB31308
Specifications
| H2AFZ | |
| Polyclonal | |
| ELISA (1:5000) Western Blot (1:1000) ChIP (0.5-1 ug/IP) Immunofluorescence (1:500) The optimal working dilution should be determined by the end user. | |
| H2A histone family, member Z | |
| H2AFZ | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of Histone HAZ. | |
| RUO | |
| 3015 | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| In PBS (0.05% sodium azide, 0.05% proclin 300). | |
| H2A.z/H2A/z/H2AZ/MGC117173 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction