Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H2AFZ, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB31310
Specifications
H2AFZ | |
Polyclonal | |
ELISA (1:500) Western Blot (1:1000) ChIP (1 ul/IP) Dot Blot (1:20000) Immunofluorescence (1:500) The optimal working dilution should be determined by the end user. | |
H2A histone family, member Z | |
H2AFZ | |
A synthetic peptide (conjugated with KLH) corresponding to Histone HAZ, acetylated at lysines 4, 7 and 11. | |
RUO | |
3015 | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
In PBS (0.05% sodium azide, 0.05% proclin 300). | |
H2A.z/H2A/z/H2AZ/MGC117173 | |
Rabbit | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction