Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H2AK119ub Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120597
This item is not returnable.
View return policy
Description
Positive test controls include: NIH/3T3, C6. Immunogen sequence: LPKKT.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications
| H2AK119ub | |
| Polyclonal | |
| Unconjugated | |
| H2ac1 | |
| H2ALys119ub; Hist1h2aa; histone cluster 1, H2aa; histone H2A type 4; Histone H2A, testis; testis-specific histone 2a; Th2a; Ubiquityl-Histone 2A K119; Ubiquityl-Histone 2A Lys119 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 24828 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.65 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q00728 | |
| H2ac1 | |
| A synthetic ubiquitinated peptide around K119 of human HIST1H2AB (NP_7344661). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction