Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2AK15ub Mouse anti-Mouse, Drosophila, Human, Clone: EDL H2AK15-4, MilliporeSigma™

Catalog No. MABE1119MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1119MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1119MI Supplier MilliporeSigma Supplier No. MABE1119
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-H2AK15ub, clone EDL H2AK15-4 is a highly specific mouse monoclonal antibody, that targets H2A ubiquitylated at Lys15 and has been tested in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H2AK15ub
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone EDL H2AK15-4
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols HIST1H2AG,HIST1H2AK
Host Species Mouse
Immunogen KLH-conjugated branched peptide corresponding to the ubiquitination site on human H2AK15.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003500.1
Test Specificity Clone EDL H2AK15-4 specifically recognizes histone H2A monoubiquitylated at Lys15, but not unmodified H2A. Does not recognize di-ubiquitin, K63 and K27 linked di-ubiquitin species.
Target Species Mouse, Drosophila, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.