Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H2BFWT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563280 Supplier Invitrogen™ Supplier No. PA563280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKEANSTTSQ KQSKQRKRGR HGPRRCHSNC RGDSFATYFR RVL Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 39%.

Atypical histone H2B. Nucleosomes containing it are structurally and dynamically indistinguishable from those containing conventional H2B. However, unlike conventional H2B, does not recruit chromosome condensation factors and does not participate in the assembly of mitotic chromosomes. May be important for telomere function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H2BFWT
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene H2BW1
Gene Accession No. Q7Z2G1
Gene Alias H2B histone family member W testis-specific; H2B histone family member W, testis specific; H2B histone family, member W, testis-specific; H2BFWT; Histone H2B type W-T
Gene Symbols H2BW1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human H2BFWT. Recombinant protein control fragment (Product #RP-100384).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.