Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H2BK11cr Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112481
Description
H2BK11cr Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transcription regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
Specifications
| H2BK11cr | |
| Polyclonal | |
| Unconjugated | |
| H2B | |
| dJ221C16.6; H2B histone family, member B; H2B.1B; H2B/a; H2B/b; H2B/g; H2B/h; H2B/k; H2B/l; H2BC10; H2BC4; H2BC6; H2BC7; H2BC8; H2BFA; H2BFB; H2BFG; H2BFH; H2BFK; H2BFL; HIRA-interacting protein 2; HIRIP2; HIST1H2BC; HIST1H2BD; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BI; histone 1, H2bd; histone cluster 1, H2bd; Histone H2B type 1-C/E/F/G/I; Histone H2B type 1-D; Histone H2B.1 A; Histone H2B.1 B; Histone H2B.a; Histone H2B.b; Histone H2B.g; Histone H2B.h; Histone H2B.k; Histone H2B.l | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3017 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P62807 | |
| H2B | |
| Peptide sequence around site of Crotonyl-Lys (11) derived from Human Histone H2B type 1-C/E/F/G/I. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction