Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H2BK20me1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112493 Supplier Invitrogen™ Supplier No. PA5112493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

H2BK20me1 Polyclonal Antibody for Western Blot, ChIP, ELISA

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transcription regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H2BK20me1
Applications ChIP Assay, ELISA, Western Blot
Classification Polyclonal
Concentration 0.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene H2B
Gene Accession No. P62807
Gene Alias dJ221C16.6; H2B histone family, member B; H2B.1B; H2B/a; H2B/b; H2B/g; H2B/h; H2B/k; H2B/l; H2BC10; H2BC4; H2BC6; H2BC7; H2BC8; H2BFA; H2BFB; H2BFG; H2BFH; H2BFK; H2BFL; HIRA-interacting protein 2; HIRIP2; HIST1H2BC; HIST1H2BD; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BI; histone 1, H2bd; histone cluster 1, H2bd; Histone H2B type 1-C/E/F/G/I; Histone H2B type 1-D; Histone H2B.1 A; Histone H2B.1 B; Histone H2B.a; Histone H2B.b; Histone H2B.g; Histone H2B.h; Histone H2B.k; Histone H2B.l
Gene Symbols H2B
Host Species Rabbit
Immunogen Peptide sequence around site of Mono-methyl-Lys (20) derived from Human Histone H2B type 1-C/E/F/G/I.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3017
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.