Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2N2 (Influenza A virus) Polyclonal Antibody, Abbomax

Catalog No. 502203959
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3959 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3959 Supplier Abbomax Inc Supplier No. 601250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: H2N2 recombinant protein

H2N2 (Influenza A virus) Polyclonal antibody specifically detects H2N2 (Influenza A virus) in Virus samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen H2N2 (Influenza A virus)
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Alias flu hemagglutinin; influenza hemagglutinin
Host Species Rabbit
Immunogen A recombinant protein of the region derived from internal region of Influenza A virus nonstructural protein (H2N2)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.