Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H3.3 G34R oncohistone mutant Recombinant Rabbit Monoclonal Antibody (RM240)
GREENER_CHOICE

Catalog No. PIMA538665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA538665 Supplier Invitrogen™ Supplier No. MA538665
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

H3.3 G34R oncohistone mutant Recombinant Monoclonal Antibody for Western Blot, IHC, IHC (P), ELISA

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H3.3 G34R oncohistone mutant
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone RM240
Concentration 250 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene H3-3A
Gene Accession No. P84243
Gene Alias H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3.3; H3.3 G34R; H3.3A; H3.3B; H3/g; H3-3A; H3-3B; H3B; H3F3; H3F3A; H3F3B; H3FT; H3t; Histone 3.3; Histone cluster 3; Histone H3; Histone H3.3; Histone H3.3 G34R; Histone H3.3G34R; PP781
Gene Symbols H3-3A, H3-3B
Host Species Rabbit
Immunogen A peptide corresponding to Histone H3.3 G34R mutant.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3020, 3021
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.