Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H3.3 K9M oncohistone mutant Recombinant Rabbit Monoclonal Antibody (22H22L9), ChIP-Verified, Invitrogen™

Catalog No. 702933
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-293-3 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-293-3 Supplier Invitrogen™ Supplier No. 702933
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Mouse, Rat. Recombinant human histone H3.3 (H3F3A, H3.3A) containing a methionine at position 9.

H3.3 K9M oncohistone mutant Recombinant Monoclonal antibody specifically detects H3.3 K9M oncohistone mutant in Human samples. It is validated for ChIP Assay, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H3.3 K9M oncohistone mutant
Applications ChIP Assay, Western Blot
Classification Recombinant Monoclonal
Clone 22H22L9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene H3-3A
Gene Accession No. P84243
Gene Alias H3 histone, family 3A; H3.3 Lys Methylated; H3.3 Lys9 M; H3.3A; H3.3B; H3.3K9M; H3-3A; H3-3B; H3F3; H3F3A; H3F3B; H3K9M; Histone H3.3; Histone H3.3 Lys9 M; PP781
Gene Symbols H3-3A
Host Species Rabbit
Immunogen Peptide corresponding to human H3.3K9M [aa6-aa14].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3020
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.