Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H3K27cr Recombinant Rabbit Monoclonal Antibody (RM401)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538669
This item is not returnable.
View return policy
Description
H3K27cr Recombinant Monoclonal Antibody for Western Blot, DB
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
Specifications
H3K27cr | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
P68431 | |
H3C1 | |
A crotonyl-peptide corresponding to Crotonyl-Histone H3 (Lys27). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Dot Blot, Western Blot | |
RM401 | |
Unconjugated | |
H3C1 | |
H3 histone family, member A; H3 K27CR; H3 Lys27 Cr; H3.1-221; H3.1-291; H3.1-I; H3/A; H3a; H3C1; H3c10; H3c11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3c8; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3g; H3h; H3i; H3Lys27Cr; Hist1h3a; HIST1H3B; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J; histone 1, H3a; histone cluster 1, H3a; Histone H3 K27 Cr; Histone H3 Lys27 Cr; histone H3.1; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; Histone H3/l | |
Rabbit | |
Protein A | |
RUO | |
8350 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction