Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H3K27me3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120811
This item is not returnable.
View return policy
Description
Positive test controls include: 293T. The target is usually found in the following locations: Chromosome, Nucleus. Immunogen sequence: KQLATKAARK SAPATGGVK.
H3K27me3 is the epigenetic mark where histone H3 is trimethylated at lysine residue 27. H3K27me3 is an epigenetic silencing mark and thus associated with downregulation of nearby genes and often found in heterochromatin regions. H3K27me3 is mainly found at inactive gene promoters and generally is found in opposition to the activating mark H3K4me3. H3K27me3 often occupies gene rich regions with close association to development regulators. EZH2, a component of the PRC2 complex, is the histone methyltransferase (or writer) responsible for trimethylating H3K27. UTX and JMJD3 both are histone demethylases (or erasers) for H3K27me3. The regulation of H3K27me3 is crucial for gene regulation and dysregulation of H3K27me3 has been implicated in numerous diseases including cancer. H3K27me3 antibodies are often used for western blot and ChIP applications to study changes in levels and occupancy of H3K27me3.
Specifications
H3K27me3 | |
Polyclonal | |
Unconjugated | |
H3C1 | |
H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3B.1; H3 histone, family 3C; H3 K27; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3A; H3.3b; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3b; h3-5; H3-53; H3-614; H3a; H3b; H3-B; H3C1; H3C10; H3c11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3c8; H3f; H3-F; H3F1K; H3F2; H3F3; h3f3a; H3f3b; h3f3b.1; H3F3C; h3f3d; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3FM; H3FN; H3g; H3h; H3i; H3k27; H3Lys27me3; h3r; Hist1; Hist1h3a; HIST1H3B; Hist1h3c; Hist1h3d; HIST1H3E; HIST1H3F; hist1h3g; hist1h3g.L; HIST1H3H; Hist1h3i; HIST1H3J; hist2h3; HIST2H3A; Hist2h3b; HIST2H3C; Hist2h3c1; Hist2h3c2; Hist2h3c2-ps; Hist2h3ca1; Hist2h3ca2; HIST2H3D; histone 1, H3a; histone 1, H3b; histone 1, H3f; histone 1, H3h; histone 2, H3a; histone 2, H3c; histone 2, H3c2; histone 2, H3ca2; Histone 3; histone cluster 1, H3a; histone cluster 1, H3b; histone cluster 1, H3f; histone cluster 1, H3g protein L homeolog; histone cluster 1, H3h; histone cluster 2, H3a; histone cluster 2, H3c; histone cluster 2, H3c2; histone cluster 2, H3c2, pseudogene; histone gene complex 1; histone H3; histone H3.1; Histone H3.2; histone H3.3; Histone H3.3C; histone H3.5; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; histone H3/l; Histone H3/m; histone H3/o; Histone H3K27me3; histone variant H3.5; hypothetical protein LOC550262; M32461; methyl Histone 3; methyl Histone H3; PP781; Trimethyl H3; Tri-methyl H3; Trimethyl Histone H3; Tri-methyl Histone H3; Tri-Methyl-H3 K27; Tri-Methyl-H3 Lys27; Tri-Methyl-Histone H3 K27; Tri-Methyl-Histone H3 Lys27; wu:fa25h06; wu:fa96g06; wu:fb07a08; wu:fb36f01; XELAEV_18028537mg; zgc:110292; zgc:174300; zgc:56193; zgc:56418; zgc:64222; zgc:86731 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
126961, 260423, 319150, 319152, 333932, 360198, 8350, 8356, 8357, 8358, 8968, 97114 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1.24 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P68431, P68433, P84228, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human histone H3 (NP_0035201). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction