Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H3K4me3Q5dopaminyl Rabbit anti-Rat, Human, Polyclonal, MilliporeSigma™

Catalog No. ABE2590100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2590100 100 μL
ABE259025UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE2590100 Supplier MilliporeSigma Supplier No. ABE2590100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

H3K4me3Q5dopaminyl Polyclonal antibody specifically detects H3K4me3Q5dopaminyl in Rat, Human samples. It is validated for Dot Blot, Peptide Inhibition Assay, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H3K4me3Q5dopaminyl
Applications Dot Blot, Inhibition Assays, Western Blot
Classification Polyclonal
Concentration 0.03 mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 10 amino acids from the N-terminal region of human Histone H3 with trimethylated lysine 4 and dopaminylated on glutamine 5.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat, Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.