Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H3K9me2/H3K14ac Recombinant Rabbit Monoclonal Antibody (RM322)
GREENER_CHOICE

Catalog No. PIMA544560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA544560 Supplier Invitrogen™ Supplier No. MA544560
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody reacts to the Histone H3 K27M mutant. No cross reactivity with wild type Histone H3.

Histone H3 is one of the five main histones involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. Histone proteins are highly post-translationally modified however Histone H3 is the most extensively modified of the five histones. The term Histone H3 alone is purposely ambiguous in that it does not distinguish between sequence variants or modification state. Histone H3 is an important protein in the emerging field of epigenetics, where its sequence variants and variable modification states are thought to play a role in the dynamic and long term regulation of genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H3K9me2/H3K14ac
Applications Western Blot, Luminex
Classification Recombinant Monoclonal
Clone RM322
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene H3-3A
Gene Accession No. P84243
Gene Symbols H3-3A, H3-3B
Host Species Rabbit
Immunogen A dimethyl-peptide corresponding to dimethyl-Histone H3 (Lys9) and an acetyl-peptide corresponding to Acetyl-Histone H3 (Lys14). The immunogen range is a synthetic peptide within to aa1-100 of histone H3 in which Lys9 is acetylated.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3020, 3021
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.