Learn More
O&M Halyard H400 Quick Check Sterilization Wrap
Catalog No. p-7225618
Click to view available options
Dimensions (L x W):
24 x 24 in.
36 x 36 in.
40 x 40 in.
45 x 45 in.
48 x 48 in.
54 x 54 in.
54 x 72 in.
This item is not returnable. View return policy
This item is not returnable. View return policy
Quick Check Sterilization Wrap is the fast, easy way to ensure instrument sterility is uncompromised.
  • Inspect trays easier and faster than ever
  • Superior microbial barrier protection with patented Powerguard technology
  • Made from recyclable #5 polypropylene fabric
  • Made in the USA
Specifications
Color Blue
Material Polypropylene
Sterility Non-sterile
Latex Free Yes
No. per Case 120/Cs.
Dimensions (L x W) 24 x 24 in.
Product Type Sterilization Wrap
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.