Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H4K8ac Recombinant Rabbit Monoclonal Antibody (RM201), ChIP-Verified
Rabbit Recombinant Monoclonal Antibody
$178.30 - $758.00
Specifications
Antigen | H4K8ac |
---|---|
Clone | RM201 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | ChIP Assay, ELISA, Western Blot, Luminex, Immunocytochemistry, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA533386
|
Invitrogen™
MA533386 |
100 μg |
Each for $758.00
|
|
|||||
PIA53338620
|
Invitrogen™
MA53338620UG |
20 μg |
Each for $178.30
|
|
|||||
Description
This product reacts to Histone H4 acetylated at Lysine 8 (K8ac). No cross reactivity with other acetylated Lysines in Histone H4. Click here for Master Lot linking information.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.Specifications
H4K8ac | |
1 mg/mL | |
ChIP Assay, ELISA, Western Blot, Luminex, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
P62805 | |
121504, 8294, 8359, 8363, 8366, 8367, 8370 | |
An acetyl-peptide corresponding to Acetyl-Histone H4 (Lys8). | |
Antibody |
RM201 | |
-20°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
H4C1 | |
acetyl Histone H4; Acetyl-H4 K8; Acetyl-H4 Lys8; Acetyl-Histone H4 K8; Acetyl-Histone H4 Lys8; CELE_T10C6.11; F07B7.11; F07B7.4; F45F2.12; FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K8ac; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/n; H4/O; H4/p; H4-12; H4-16; H4-53; H4c1; H4c11; H4C12; H4C13; H4C14; H4C15; H4c2; H4c3; H4C4; H4C5; H4c6; H4C8; H4c9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K8; H4Lys8ac; H4M; his-20; his-22; his-4; his-52; his-54; his-8; HIST1H4; HIST1H4A; Hist1h4b; HIST1H4C; Hist1h4d; HIST1H4E; HIST1H4F; HIST1H4H; Hist1h4i; Hist1h4j; HIST1H4K; HIST1H4L; Hist1h4m; Hist2h4; HIST2H4A; HIST2H4B; Hist4h4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; Histone H2B 2; Histone H4; Histone H4 acetyl; Histone H4A; Histone H4K8; Histone H4K8ac; histone IV, family 2; K06C4.12; K06C4.4; T10C6.11; X04652 | |
H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9 | |
Primary | |
Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title