Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H4K91ac Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5105104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105104 Supplier Thermo Scientific Supplier No. PA5105104

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Acetyl-Histone H4 only when acetylated at Lys91.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H4K91ac
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene H4C1
Gene Accession No. P62803, P62805, P62806
Gene Alias acetyl Histone H4; Acetyl-H4 K91; Acetyl-H4 Lys91; Acetyl-Histone H4 K91; Acetyl-Histone H4 Lys91; FO108; H4; H4 clustered histone 2; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K91ac; H4.1; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/n; H4/O; H4/p; H4-12; H4-16; H4-53; H4C1; H4c11; H4C12; H4C13; H4C14; H4C15; H4C2; H4C3; H4C4; H4C5; H4c6; H4c8; H4c9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K91; H4Lys91ac; H4M; HIST1H4; Hist1h4a; Hist1h4b; Hist1h4c; Hist1h4d; HIST1H4E; HIST1H4F; Hist1h4h; HIST1H4I; Hist1h4j; HIST1H4K; HIST1H4L; Hist1h4m; Hist2h4; Hist2h4a; HIST2H4B; HIST4H4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; histone H3; histone H4; Histone H4 acetyl; histone H4.1; Histone H4A; Histone H4K91; Histone H4K91ac; histone IV, family 2; LOC102161782; X04652
Gene Symbols H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9, LOC102161782
Host Species Rabbit
Immunogen A synthesized peptide derived from human Histone H4(Accession P62805), corresponding to amino acid residues R79-G100.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102161782, 121504, 280691, 319158, 326619, 326620, 8294, 8359, 8363, 8366, 8367, 8370, 97122
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.