Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ H5N8 (A/duck/NY/191255-59/02, AAP72011, 1 a.a. - 490 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells

Catalog No. 89947005
Click to view available options
Quantity:
50 μg

Used for SDS-PAGE

Specifications

Accession Number AAP72011
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Name H5N8 (A/duck/NY/191255-59/02) Recombinant Protein
Preparation Method Mammalian cell (293) expression system
Quantity 50 μg
Storage Requirements Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name H5N8
Species Human
Recombinant Recombinant
Protein Tag His
Expression System Mammalian cell (293) expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.