Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H60 Antibody (205326), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1155K
This item is not returnable.
View return policy
Description
H60 Monoclonal antibody specifically detects H60 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA (Matched Antibody Pair)Specifications
H60 | |
Monoclonal | |
DyLight 488 | |
H60a | |
BaF3 mouse pro-B cell line transfected with mouse H60 and mouse myeloma cell line NS0-derived recombinant mouse H60, Asp30-Gln212, Accession # Q3TDZ7 | |
0.5 mL | |
Cancer | |
15101 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, ELISA | |
205326 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction