Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H60 Antibody (RM0248-11G35), Alexa Fluor™ 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212428AF488
Description
H60 Monoclonal specifically detects H60 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
H60 | |
Monoclonal | |
Alexa Fluor 488 | |
H60a | |
Mouse recombinant protein of H60. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry | |
RM0248-11G35 | |
Western Blot, Flow Cytometry | |
Rat | |
Protein A or G purified | |
RUO | |
15101 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction