Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H60 Antibody (RM0248-11G35), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212428MFV610
Description
H60 Monoclonal antibody specifically detects H60 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
H60 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
RM0248-11G35 | |
Flow Cytometry | |
H60a | |
Mouse recombinant protein of H60. | |
0.1 mL | |
Cancer | |
15101 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction