Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H60 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547081
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Enables natural killer cell lectin-like receptor binding activity. Acts upstream of or within several processes, including positive regulation of immune response; positive regulation of interferon-gamma production; and positive regulation of leukocyte activation.
Specifications
| H60 | |
| Polyclonal | |
| Unconjugated | |
| H60a | |
| H60; H60a; histocompatibility 60; histocompatibility 60a | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 15101 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| 0 | |
| H60a | |
| Mouse myeloma cell line NS0-derived recombinant mouse H60 Asp30-Gln212. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction