Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H6PD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592977 Supplier Invitrogen™ Supplier No. PA592977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QELQGHVSII LLGATGDLAK KYLWQGLFQL YLDEAGRGHS FSFHGAALTA PKQGQELMAK ALESLSCPKD MAPSHCAEHK DQFLQLSQYR QLKTAEDYQA LNKDIEAQLQ HAGLREAGRI FYFSVPPFAY EDIARNINSS CRPGPGAWLR VVLEKPFGHD HFSAQQLATE LGTFFQEEEM Y; Positive Samples: Mouse liver, Mouse kidney; Cellular Location: Endoplasmic reticulum lumen.

The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a type 2 salivary cysteine peptidase inhibitor. The protein is an S-type cystatin, based on its high level of expression in saliva, tears and seminal plasma. The specific role in these fluids is unclear but antibacterial and antiviral activity is present, consistent with a protective function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H6PD
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene H6PD
Gene Accession No. O95479, Q8CFX1
Gene Alias 6PGL; 6-phosphogluconolactonase; AI785303; CORTRD1; EGK_00198; G6PD, H form; G6pd1; G6PDH; GDH; GDH/6PGL endoplasmic bifunctional protein; glucose 1- dehydrogenase; glucose 1-dehydrogenase; glucose dehydrogenase; glucose dehyrogenase; Glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase 1; glucose-6-phosphate dehydrogenase, salivary; Gpd1; Gpd-1; H6pd; H6pdh; hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase); hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase
Gene Symbols H6PD
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-200 of human H6PD (NP_0042762).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100198, 298655, 9563
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.